Ethylenediaminetetraacetic acid disodium nickel(II) salt hydrate

Proveedor: TCI
Peligro

E0097-25G
TCIAE0097-25GEA 236 EUR
TCIAE0097-25G
Ethylenediaminetetraacetic acid disodium nickel(II) salt hydrate
Ethylenediaminetetraacetic acid disodium nickel(II) salt hydrate
Formula: C₁₀H₁₆N₂O₈
Número MDL: MFCD00070493
Núm. CAS: 15708-55-1

Order Now

Resultados test especificaciones

Appearance Blue to Dark blue to Dark purple powder to crystal
Purity (Chelometric Titration) min. 98.0 % (calcd.on anh.substance)
Solubility in Water almost transparency
Water max. 17.0 %

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR