Potassium-2-ethylhexanoate ≥95.0% (by titrimetric analysis)

Proveedor: TCI
Atención

E0121-25G E0121-500G
TCIAE0121-25GEA 41 EUR
TCIAE0121-25G TCIAE0121-500G
Potassium-2-ethylhexanoate ≥95.0% (by titrimetric analysis)
Potassium-2-ethylhexanoate
Formula: C₈H₁₆O₂
Número MDL: MFCD00045896
Núm. CAS: 3164-85-0
EINECS: 221-625-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=863f3de8ca91712ab8c53834ff8af279c9551d57&vid=bbb61511e7403dbb983c501c96093ac7f555a7f8&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White or Colorless powder to lump
Purity (Nonaqueous Titration) min. 95.0 %
Water max. 5.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR