Ethyl cellulose, 9-11 mPa.s [5% in Toluene + Ethanol (80:20), 25 °C]

Proveedor: TCI

Sinónimos: Ethylcellulose

E0265-500G E0265-25G
TCIAE0265-500GEA 111 EUR
TCIAE0265-500G TCIAE0265-25G
Ethyl cellulose, 9-11 mPa.s [5% in Toluene + Ethanol (80:20), 25 °C]
Ethyl cellulose
Número MDL: MFCD00131037
Núm. CAS: 9004-57-3
Índice Merck: 14,03781

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Solubility in Toluene-EtOH mix. almost transparency
Drying loss max. 2.0 %
Viscosity 9.0 to 11.0 mPa-s (5 %, Toluene:EtOH=80:20, 25 deg-C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR