Ethyl (R)-4-phenyl-2-(trifluoromethanesulphonyloxy)butyrate ≥97.0% (por GC)

Proveedor: TCI

E0879-1G
TCIAE0879-1GEA 131 EUR
TCIAE0879-1G
Ethyl (R)-4-phenyl-2-(trifluoromethanesulphonyloxy)butyrate ≥97.0% (por GC)
Ethyl (R)-4-phenyl-2-(trifluoromethanesulphonyloxy)butyrate
Formula: C₁₃H₁₅F₃O₅S
Número MDL: MFCD09751096
Núm. CAS: 88767-98-0

Order Now

Resultados test especificaciones

Appearance Light yellow to Amber to Dark green clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR