L-Glutamic acid 5-ethyl ester ≥97.0% (by titrimetric analysis)

Proveedor: TCI

Sinónimos: 5-Ethyl L-Glutamate, L-Glutamic acid 5-Ethyl ester, H-Glu(OEt)-OH

E1155-5G E1155-25G
TCIAE1155-5GEA 22 EUR
TCIAE1155-5G TCIAE1155-25G
L-Glutamic acid 5-ethyl ester ≥97.0% (by titrimetric analysis)
L-Glutamic acid 5-ethyl ester
Formula: C₇H₁₃NO₄
Temperatura de almacenaje: Nevera
Número MDL: MFCD00037217
Núm. CAS: 1119-33-1
EINECS: 214-274-6
Índice Merck: 14,04470

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 97.0 %
Specific rotation [a]20/D +11.0 to +14.0 deg (C=1,H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR