Ethyl 2-(3-Formyl-4-hydroxyphenyl)-4-methylthiazole-5-carboxylate ≥98.0% (by GC, titration analysis)

Proveedor: TCI

E1394-5G E1394-25G
TCIAE1394-5GEA 30 EUR
TCIAE1394-5G TCIAE1394-25G
Ethyl 2-(3-Formyl-4-hydroxyphenyl)-4-methylthiazole-5-carboxylate ≥98.0% (by GC, titration analysis)
Ethyl 2-(3-Formyl-4-hydroxyphenyl)-4-methylthiazole-5-carboxylate
Formula: C₁₄H₁₃NO₄S
Número MDL: MFCD13194810
Núm. CAS: 161798-01-2

Order Now

Resultados test especificaciones

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 113.0 to 117.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR