2-Fluorofenilhidracinio cloruro ≥97.0% (by titrimetric analysis)

Proveedor: TCI
Atención

F0368-5G F0368-1G
TCIAF0368-5GEA 49 EUR
TCIAF0368-5G TCIAF0368-1G
2-Fluorofenilhidracinio cloruro ≥97.0% (by titrimetric analysis)
2-Fluorofenilhidracinio cloruro
Formula: C₆H₈ClFN₂
Número MDL: MFCD00012927
Núm. CAS: 2924-15-4
EINECS: 220-885-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=051f19a5dd86671f060a04b47a9316821bdbdbc1&vid=051f19a5dd86671f060a04b47a9316821bdbdbc1&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light orange powder to crystal
Purity (Potassium iodate Method) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR