Fmoc-D-Glu-OH ≥97.0% (by HPLC, titration analysis)

Proveedor: TCI

F0600-25G F0600-5G
TCIAF0600-25GEA 105 EUR
TCIAF0600-25G TCIAF0600-5G
Fmoc-D-Glu-OH ≥97.0% (by HPLC, titration analysis)
Fmoc-D-Glu-OH
Formula: C₂₀H₁₉NO₆
Número MDL: MFCD09037362
Núm. CAS: 104091-09-0

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 97.0 %
Specific rotation [a]20/D +18.0 to +22.0 deg (C=1, DMF)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR