5-Fluoro-3-pyridineboronic acid (contains varying amounts of Anhydride)

Proveedor: TCI
Atención

F0859-1G F0859-5G
TCIAF0859-1GEA 71 EUR
TCIAF0859-1G TCIAF0859-5G
5-Fluoro-3-pyridineboronic acid (contains varying amounts of Anhydride)
5-Fluoro-3-pyridineboronic acid
Formula: C₅H₅BFNO₂
Número MDL: MFCD07368243
Núm. CAS: 872041-86-6

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 115.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR