2-Fluoropiridin-3-ol ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

F0886-1G F0886-5G
TCIAF0886-1GEA 50 EUR
TCIAF0886-1G TCIAF0886-5G
2-Fluoropiridin-3-ol ≥98.0% (by GC, titration analysis)
2-Fluoropiridin-3-ol
Formula: C₅H₄FNO
Número MDL: MFCD04112569
Núm. CAS: 174669-74-0

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 126.0 to 131.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR