N-Glycyl-DL-2-aminobutyric acid

Proveedor: TCI

G0114-1G
TCIAG0114-1GEA 47 EUR
TCIAG0114-1G
N-Glycyl-DL-2-aminobutyric acid
Proteínas y Péptidos

  • Melting point: 230 °C
  • Solubility in wtaer: almost transparency

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR