Calcium-DL-glycerate hydrate ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Atención

Sinónimos: Calcium 2,3-Dihydroxypropionate, DL-Glyceric Acid Calcium Salt, 2,3-Dihydroxypropionic Acid Calcium Salt

G0232-10G
TCIAG0232-10GEA 197 EUR
TCIAG0232-10G
Calcium-DL-glycerate hydrate ≥98.0% (by titrimetric analysis)
Calcium-DL-glycerate hydrate
Formula: C₃H₆O₄
Número MDL: MFCD00149289
Núm. CAS: 67525-74-0

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 98.0 % (calcd.on anh.substance)
Solubility in hot Water almost transparency
Water max. 15.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR