Graphene, nanoplatelets, Length: 5 µm, Ø 6-8 nm

Proveedor: TCI
Atención

G0441-5G G0441-25G
TCIAG0441-5GEA 56 EUR
TCIAG0441-5G TCIAG0441-25G
Graphene, nanoplatelets, Length: 5 µm, Ø 6-8 nm
Graphene
Número MDL: MFCD00144065
Núm. CAS: 1034343-98-0

Order Now

Resultados test especificaciones

Appearance Black powder to lump
Thick 6 to 8 nm
Average size Equal to 5 micro_m

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR