Hydrazine-L-(+)-tartrate ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Peligro

H0175-25G
TCIAH0175-25GEA 91 EUR
TCIAH0175-25G
Hydrazine-L-(+)-tartrate ≥98.0% (by titrimetric analysis)
Hydrazine-L-(+)-tartrate
Formula: H₄N₂
MW: 182.13 g/mol
Número MDL: MFCD00070610
Núm. CAS: 634-62-8
EINECS: 211-211-4
UN: 2811
ADR: 6.1 III

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Potassium iodate Method) min. 98.0 %
Specific rotation [a]20/D +22.0 to +25.0 deg (C=1, H2O)
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR