Sodium-1-hexadecanesulphonate

Proveedor: TCI

H0415-10G H0415-25G
TCIAH0415-10GEA 54 EUR
TCIAH0415-10G TCIAH0415-25G
Sodium-1-hexadecanesulphonate
Sodium-1-hexadecanesulphonate
Formula: C₁₆H₃₃NaSO₃
Número MDL: MFCD00007528
Núm. CAS: 15015-81-3
EINECS: 239-100-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=63cc5535bb7b50e1f0f3fc443f57cfc168a7ecb9&vid=63cc5535bb7b50e1f0f3fc443f57cfc168a7ecb9&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Solubility in hot Water almost transparency
Drying loss max. 2.0 %
Ignition residue (Sulfate) 20.8 to 21.9 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR