Hidroxipropilcelulosa, 1,000-4,000 mPa.s (2% in H₂O 20 °C)

Proveedor: TCI

H0475-25G H0475-500G
TCIAH0475-25GEA 24 EUR
TCIAH0475-25G TCIAH0475-500G
Hidroxipropilcelulosa, 1,000-4,000 mPa.s (2% in H₂O 20 °C)
Hydroxypropyl cellulose
Número MDL: MFCD00132688
Núm. CAS: 9004-64-2
Índice Merck: 14,04841

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Solubility in Water almost transparency
Drying loss max. 5.0 %
Viscosity 1,000 to 4,000 mPa-s (2wt%, H2O, 20 deg-C)

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR