1,5-Heptadiene (cis and trans mixture) cis- and trans-mixture ≥97.0% (por GC)

Proveedor: TCI
Peligro

H0551-5ML
TCIAH0551-5MLEA 125 EUR
TCIAH0551-5ML
1,5-Heptadiene (cis and trans mixture) cis- and trans-mixture ≥97.0% (por GC)
1,5-Heptadiene (cis and trans mixture)
Formula: C₇H₁₂
MW: 96.17 g/mol
Pto. de inflamación: –3°C
Número MDL: MFCD00026998
Núm. CAS: 1541-23-7
UN: 3295
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8774268b9f25d6dd721edf6530fa479179c61470&vid=a26308cff7c043da3c879d1a3fd95296397ac2a1&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR