3-Hydroxy-o-toluic acid ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Atención

H0885-1G H0885-25G H0885-5G
TCIAH0885-1GEA 22 EUR
TCIAH0885-1G TCIAH0885-25G TCIAH0885-5G
3-Hydroxy-o-toluic acid ≥98.0% (by titrimetric analysis)
3-Hydroxy-o-toluic acid
Formula: C₈H₈O₃
Número MDL: MFCD00671541
Núm. CAS: 603-80-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=33447f7ebfd70954ca84ea91bb4e34158843b29b&vid=33447f7ebfd70954ca84ea91bb4e34158843b29b&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 143.0 to 147.0 deg-C
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR