6-Hydroxypyrimidine-4-carboxylic acid ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI

H1392-200MG H1392-1G
TCIAH1392-200MGEA 28 EUR
TCIAH1392-200MG TCIAH1392-1G
6-Hydroxypyrimidine-4-carboxylic acid ≥98.0% (by HPLC, titration analysis)
6-Hydroxypyrimidine-4-carboxylic acid
Formula: C₅H₄N₂O₃
Número MDL: MFCD00023267
Núm. CAS: 6299-87-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=63044b12b6e122eab5def70fe332568240b0d5f8&vid=63044b12b6e122eab5def70fe332568240b0d5f8&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light red powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR