(2-Hydroxy-5-methylphenyl)triphenylphosphonium bromide ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

H1748-1G H1748-5G
TCIAH1748-1GEA 50 EUR
TCIAH1748-1G TCIAH1748-5G
(2-Hydroxy-5-methylphenyl)triphenylphosphonium bromide ≥98.0% (by HPLC, titration analysis)
(2-Hydroxy-5-methylphenyl)triphenylphosphonium bromide
Formula: C₂₅H₂₂BrOP
Núm. CAS: 2005487-65-8

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystaline
Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 98.0 %

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR