Isobutylamine hydrochloride ≥99.0% (by titrimetric analysis)

Proveedor: TCI
Atención

Sinónimos: 2-Methylpropan-1-amine hydrochloride, 1-Amino-2-methylpropane Hydrochloride

I0096-25G I0096-500G
TCIAI0096-25GEA 28 EUR
TCIAI0096-25G TCIAI0096-500G
Isobutylamine hydrochloride ≥99.0% (by titrimetric analysis)
Isobutylamine hydrochloride
Formula: C₄H₁₁N
MW: 109.60 g/mol
Número MDL: MFCD00060199
Núm. CAS: 5041-09-8

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 99.0 %
Purity (Argentmetric Titration) min. 99.0 %
Melting point 176.0 to 180.0 deg-C
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR