Isoniazida ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 4-Picolinic acid hydrazide, 4-Pyridinecarboxylic acid hydrazide, Isonicotinic hydrazide, Pyridine-4-carboxylic acid hydrazide, Isonicotinic acid hydrazide

I0138-25G I0138-500G I0138-100G
TCIAI0138-25GEA 21 EUR
TCIAI0138-25G TCIAI0138-500G TCIAI0138-100G
Isoniazida ≥98.0% (by HPLC, titration analysis)
Isoniazida
Formula: C₆H₇N₃O
MW: 137.14 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00006426
Núm. CAS: 54-85-3
EINECS: 200-214-6
Índice Merck: 14,05186
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1946f0c3a3d86d9b196f289e965ec92a006cfcb9&vid=1946f0c3a3d86d9b196f289e965ec92a006cfcb9&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Water almost transparency
Melting point 170.0 to 173.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR