3-(Hidroximetil)indol ≥98.0% (by HPLC, total nitrogen)

Proveedor: TCI

I0496-25G I0496-5G
TCIAI0496-25GEA 141 EUR
TCIAI0496-25G TCIAI0496-5G
3-(Hidroximetil)indol ≥98.0% (by HPLC, total nitrogen)
3-(Hidroximetil)indol
Formula: C₉H₉NO
Temperatura de almacenaje: Congelador
Número MDL: MFCD00005632
Núm. CAS: 700-06-1
EINECS: 211-836-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32adee6338a022600954052326beb972e80b8139&vid=32adee6338a022600954052326beb972e80b8139&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR