Itopride hydrochloride ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI

I0948-1G I0948-5G I0948-25G
TCIAI0948-1GEA 16 EUR
TCIAI0948-1G TCIAI0948-5G TCIAI0948-25G
Itopride hydrochloride ≥98.0% (by HPLC, titration analysis)
Itopride hydrochloride
Formula: C₂₀H₂₆N₂O₄·HCl
Temperatura de almacenaje: Nevera
Número MDL: MFCD00881710
Núm. CAS: 122892-31-3
Índice Merck: 14,05244
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=59d6d06aa43b5d0e353a96372052d6f52b2f6cd6&vid=59d6d06aa43b5d0e353a96372052d6f52b2f6cd6&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 192.0 to 196.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR