Isopentylamine hydroiodide ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Atención

I1095-1G I1095-5G
TCIAI1095-1GEA 26 EUR
TCIAI1095-1G TCIAI1095-5G
Isopentylamine hydroiodide ≥98.0% (by titrimetric analysis)
Isopentylamine hydroiodide
Formula: C₅H₁₃N·HI
Número MDL: MFCD33021473
Núm. CAS: 2733412-76-3

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR