DL-Leucic acid ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

L0104-25G L0104-500G
TCIAL0104-25GEA 101 EUR
TCIAL0104-25G TCIAL0104-500G
DL-Leucic acid ≥98.0% (by GC, titration analysis)
DL-Leucic acid
Formula: C₆H₁₂O₃
Número MDL: MFCD00004246
Núm. CAS: 498-36-2
EINECS: 207-860-8

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 74.0 to 78.0 deg-C
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR