L-Leucine ethyl ester hydrochloride ≥99.0% (by titrimetric analysis)

Proveedor: TCI

L0123-25G L0123-5G
TCIAL0123-25GEA 67 EUR
TCIAL0123-25G TCIAL0123-5G
L-Leucine ethyl ester hydrochloride ≥99.0% (by titrimetric analysis)
L-Leucine ethyl ester hydrochloride
Formula: C₈H₁₇NO₂
Número MDL: MFCD00034879
Núm. CAS: 2743-40-0
EINECS: 220-375-6

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Argentmetric Titration) min. 99.0 %
Specific rotation [a]20/D +18.0 to +20.0 deg (C=5, EtOH)
Solubility in Water almost transparency

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR