D-Leucine benzyl ester-p-toluenesulphonate ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI

L0194-5G
TCIAL0194-5GEA 60 EUR
TCIAL0194-5G
D-Leucine benzyl ester-p-toluenesulphonate ≥98.0% (by HPLC, titration analysis)
D-Leucine benzyl ester-p-toluenesulphonate
Formula: C₁₃H₁₉NO₂
Número MDL: MFCD00066116
Núm. CAS: 17664-93-6

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 % (total of p-TS)
Purity (Neutralization titration) min. 98.0 %
Melting point 156.0 to 160.0 deg-C
Specific rotation [a]20/D -3.0 to -6.0 deg (C=2, DMF)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR