Lisinopril dihydrate ≥98.0% (by titrimetric analysis)

Proveedor: TCI

L0220-1G L0220-5G
TCIAL0220-1GEA 42 EUR
TCIAL0220-1G TCIAL0220-5G
Lisinopril dihydrate ≥98.0% (by titrimetric analysis)
Lisinopril dihydrate
Formula: C₂₁H₃₅N₃O₇
Pto. de fusión: 148°C
Temperatura de almacenaje: Nevera
Número MDL: MFCD01698825
Núm. CAS: 83915-83-7
Índice Merck: 14,05516
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9ef3dc83ff5a9dc83d5c4121184184294743ea64&vid=9ef3dc83ff5a9dc83d5c4121184184294743ea64&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D -42.0 to -48.0 deg (C=1, 0.25mol/L Zinc Acetate buffer sol.) (calcd.on anh.substance)
Water 8.0 to 9.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR