Lumefantrine ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI

L0256-5G L0256-25G
TCIAL0256-5GEA 70 EUR
TCIAL0256-5G TCIAL0256-25G
Lumefantrine ≥98.0% (by HPLC, titration analysis)
Lumefantrine
Formula: C₃₀H₃₂Cl₃NO
Número MDL: MFCD05662268
Núm. CAS: 82186-77-4
Índice Merck: 14,05597
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5d348452b280f062716c57cda65bdd2099349be7&vid=5d348452b280f062716c57cda65bdd2099349be7&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Light yellow to Amber to Dark green powder to crystaline
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 134.0 to 138.0 deg-C

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR