Lithium diphenylphosphide ∼0.5 mol/L in THF

Proveedor: TCI
Peligro

L0302-25ML
TCIAL0302-25MLEA 177 EUR
TCIAL0302-25ML
Lithium diphenylphosphide ∼0.5 mol/L in THF
Lithium diphenylphosphide
Formula: C₁₂H₁₀LiP
Número MDL: MFCD00046054
Núm. CAS: 4541-02-0
UN: 2924
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c15b5178e34d770f849b3e9948659dcb2f7f3697&vid=c15b5178e34d770f849b3e9948659dcb2f7f3697&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Concentration (Nonaqueous Titration) 9.5 to 11.3 w/w%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR