β-Methyllevulinic acid ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

M1057-25G M1057-5G
TCIAM1057-25GEA 795 EUR
TCIAM1057-25G TCIAM1057-5G
β-Methyllevulinic acid ≥98.0% (by GC, titration analysis)
beta-Methyllevulinic acid
Formula: C₆H₁₀O₃
Pto. Ebullición: 242°C
Temperatura de almacenaje: Nevera
Número MDL: MFCD00143691
Núm. CAS: 6628-79-1

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 32.0 to 35.0 deg-C
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR