5-Metiltetrazol ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Peligro

M1069-25G
TCIAM1069-25GEA 106 EUR
TCIAM1069-25G
5-Metiltetrazol ≥98.0% (by HPLC, titration analysis)
5-Metiltetrazol
Formula: C₂H₄N₄
Número MDL: MFCD00129971
Núm. CAS: 4076-36-2
UN: 1325
ADR: 4.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5630aca9a11b5ef1b8a655c991f26d90e1cf2723&vid=5630aca9a11b5ef1b8a655c991f26d90e1cf2723&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 145.0 to 148.0 deg-C
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR