Bis(4-nitrobenzyl)malonate ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: Malonic Acid Bis(4-nitrobenzyl) Ester

M1181-5G
TCIAM1181-5GEA 107 EUR
TCIAM1181-5G
Bis(4-nitrobenzyl)malonate ≥98.0% (by HPLC, titration analysis)
Bis(4-nitrobenzyl)malonate
Formula: C₁₇H₁₄N₂O₈
MW: 374,31 g/mol
Número MDL: MFCD00191671
Núm. CAS: 67245-85-6

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Ester Value) min. 98.0 %
Melting point 87.0 to 91.0 deg-C
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR