Methylaluminum bis(2,6-di-tert-butyl-4-methylphenoxide) 0.4 mol/L in toluene

Proveedor: TCI
Peligro

Sinónimos: Bis(2,6-di-tert-butyl-4-methylphenoxy)methylaluminum

M1211-50ML
TCIAM1211-50MLEA 96 EUR
TCIAM1211-50ML
Methylaluminum bis(2,6-di-tert-butyl-4-methylphenoxide) 0.4 mol/L in toluene
Methylaluminum bis(2,6-di-tert-butyl-4-methylphenoxide)
Formula: C₃₁H₄₉AlO₂
MW: 480,71 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD00079652
Núm. CAS: 56252-55-2
UN: 3399
ADR: 4.3,II

Order Now

Resultados test especificaciones

Appearance Colorless to Red to Green clear liquid
Concentration (Chelometric Method)unit2 19.0 to 24.0 w/w%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR