L-Glutamic acid 5-methyl ester ≥98.0% (by titrimetric analysis)

Proveedor: TCI

Sinónimos: H-Glu(OMe)-OH

M1690-25G M1690-5G
TCIAM1690-25GEA 89 EUR
TCIAM1690-25G TCIAM1690-5G
L-Glutamic acid 5-methyl ester ≥98.0% (by titrimetric analysis)
L-Glutamic acid 5-methyl ester
Formula: C₆H₁₁NO₄
Número MDL: MFCD00002632
Núm. CAS: 1499-55-4
EINECS: 216-110-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=76979773a6330a20571d5762f2a4720501fe5a06&vid=76979773a6330a20571d5762f2a4720501fe5a06&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +28.0 to +30.0 deg (C=2, 6mol/L HCl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR