Methylamine hydrobromide, low water content ≥98.0% (by total nitrogen and titration analysis)

Proveedor: TCI
Atención

M2589-5G M2589-1G M2589-25G
TCIAM2589-5GEA 128 EUR
TCIAM2589-5G TCIAM2589-1G TCIAM2589-25G
Methylamine hydrobromide, low water content ≥98.0% (by total nitrogen and titration analysis)
Methylamine hydrobromide
Formula: CH₅N·HBr
Núm. CAS: 6876-37-5
EINECS: 229-981-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a4a77f33be7281b4bbc5bc18b32be3ea67a91774&vid=a4a77f33be7281b4bbc5bc18b32be3ea67a91774&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point 248.0 to 253.0 deg-C
Water max. 100 ppm

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR