Magnesiumbis(trifluoromethylsulphonyl)imide ≥97.0% (by titrimetric analysis)

Proveedor: TCI
Peligro

M2861-1G M2861-5G
TCIAM2861-1GEA 43 EUR
TCIAM2861-1G TCIAM2861-5G
Magnesiumbis(trifluoromethylsulphonyl)imide ≥97.0% (by titrimetric analysis)
Magnesiumbis(trifluoromethylsulphonyl)imide
Formula: C₄F₁₂MgN₂O₈S₄
Número MDL: MFCD14156020
Núm. CAS: 133395-16-1
UN: 1759
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af4df9c6112b31fd2244563debeba0a3a1251b6f&vid=af4df9c6112b31fd2244563debeba0a3a1251b6f&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 97.0 %
Purity (Ion exchange titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR