Boc-Asp-OMe ≥97.0% (by HPLC, titration analysis)

Proveedor: TCI

Sinónimos: Boc-Asp-OMe, N-Boc-L-aspartic acid 1-Methyl ester, N-Boc-L-aspartic acid 1-methyl ester, 1-Methyl N-Boc-L-aspartate, 1-Methyl N-(tert-Butoxycarbonyl)-L-aspartate, N-(tert-Butoxycarbonyl)-L-aspartic acid 1-Methyl ester

M2970-5G
TCIAM2970-5GEA 56 EUR
TCIAM2970-5G
Boc-Asp-OMe ≥97.0% (by HPLC, titration analysis)
Boc-Asp-OMe
Formula: C₁₀H₁₇NO₆
MW: 247.247 g/mol
Número MDL: MFCD03094777
Núm. CAS: 98045-03-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=17f4bb97b317f5b874f4abd4d81f49afa59490c3&vid=17f4bb97b317f5b874f4abd4d81f49afa59490c3&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 97.0 %
Melting point 87.0 to 91.0 deg-C
Specific rotation [a]20/D -20.0 to -23.0 deg (C=1, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR