2-Methoxy-N-(4-methoxyphenyl)aniline ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

M3067-1G M3067-5G
TCIAM3067-1GEA 142 EUR
TCIAM3067-1G TCIAM3067-5G
2-Methoxy-N-(4-methoxyphenyl)aniline ≥98.0% (by GC, titration analysis)
2-Methoxy-N-(4-methoxyphenyl)aniline
Formula: C₁₄H₁₅NO₂
Número MDL: MFCD27956479
Núm. CAS: 58751-07-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=439986587f627f53cf047b4574e6e5f029c0612b&vid=439986587f627f53cf047b4574e6e5f029c0612b&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 69.0 to 73.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR