Methyl 2-Amino-5-dodecylbenzoate ≥96.0% (por GC)

Proveedor: TCI
Atención

Sinónimos: 2-Amino-5-dodecylbenzoic acid methyl ester

M3234-1G
TCIAM3234-1GEA 124 EUR
TCIAM3234-1G
Methyl 2-Amino-5-dodecylbenzoate ≥96.0% (por GC)
Methyl 2-Amino-5-dodecylbenzoate
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=18cfa2661ffaf8edf7a47beb8957e6b48823b3e1&vid=de1f6ec1075748f9d5714d1911fc1eca694c18ff&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 96.0 %
Melting point 52.0 to 56.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR