Sodium-2-naphthol-7-sulphonate hydrate ≥98.0% (by titrimetric analysis)

Proveedor: TCI

N0036-25G
TCIAN0036-25GEA 60 EUR
TCIAN0036-25G
Sodium-2-naphthol-7-sulphonate hydrate ≥98.0% (by titrimetric analysis)
Sodium-2-naphthol-7-sulphonate hydrate
Formula: C₁₀H₈O₄S
Número MDL: MFCD00021625
Núm. CAS: 135-55-7
EINECS: 205-200-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=18f9bf747d9e19ead52be22c77ec338679a25594&vid=18f9bf747d9e19ead52be22c77ec338679a25594&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Gray to Red powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (Ion exchange titration) min. 98.0 % (calcd.on anh.substance) (after deduct NaCl content)
Water max. 8.0 %

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR