Perfluorobutano-1-sulfonato de potasio ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Atención

N0711-25G
TCIAN0711-25GEA 63 EUR
TCIAN0711-25G
Perfluorobutano-1-sulfonato de potasio ≥98.0% (by titrimetric analysis)
Perfluorobutano-1-sulfonato de potasio
Formula: C₄F₉KO₃S
Número MDL: MFCD00007515
Núm. CAS: 29420-49-3
EINECS: 249-616-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=551c6f00aa6d1286be7a4a57af6215001c19a09d&vid=551c6f00aa6d1286be7a4a57af6215001c19a09d&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Ion exchange titration) min. 98.0 %
Solubility in Methanol almost transparency

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR