4-(1-Naphthyl)phenylboronic acid (contains varying amounts of Anhydride)

Proveedor: TCI
Atención

N0798-1G N0798-5G
TCIAN0798-1GEA 33 EUR
TCIAN0798-1G TCIAN0798-5G
4-(1-Naphthyl)phenylboronic acid (contains varying amounts of Anhydride)
4-(1-Naphthyl)phenylboronic acid
Formula: C₁₆H₁₃BO₂
Número MDL: MFCD08669639
Núm. CAS: 870774-25-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7933fb0b85c159d95e7a64dc243b0542d445ffe2&vid=7933fb0b85c159d95e7a64dc243b0542d445ffe2&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) 97.0 to 108.0 %

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR