Phenyl phosphate disodium salt ≥98.0% (by titrimetric analysis)

Proveedor: TCI

P0208-25G P0208-500G
TCIAP0208-25GEA 55 EUR
TCIAP0208-25G TCIAP0208-500G
Phenyl phosphate disodium salt ≥98.0% (by titrimetric analysis)
Phenyl phosphate disodium salt
Formula: C₆H₅Na₂O₄P·xH₂O
Número MDL: MFCD00002133
Núm. CAS: 3279-54-7
EINECS: 221-917-4
Índice Merck: 14,03357

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 % (calcd.on anh.substance)
Solubility in Water almost transparency
Water 7.0 to 16.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR