9,10-Bis(phenylethynyl)anthracene ≥98.0% (by HPLC)

Proveedor: TCI
Atención

P1137-1G P1137-5G
TCIAP1137-1GEA 26 EUR
TCIAP1137-1G TCIAP1137-5G
9,10-Bis(phenylethynyl)anthracene ≥98.0% (by HPLC)
9,10-Bis(phenylethynyl)anthracene
Formula: C₃₀H₁₈
Número MDL: MFCD00012050
Núm. CAS: 10075-85-1
EINECS: 233-210-8
Índice Merck: 14,01299
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=042859c24942c73a2a922d2f84f7c23c42f0ed61&vid=042859c24942c73a2a922d2f84f7c23c42f0ed61&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Orange to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Chemiluminescence test to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR