Tetrabenzyl pyrophosphate ≥98.0% (by HPLC)

Proveedor: TCI
Peligro

Sinónimos: Tetrabenzyl diphosphate

P1223-1G
TCIAP1223-1GEA 75 EUR
TCIAP1223-1G
Tetrabenzyl pyrophosphate ≥98.0% (by HPLC)
Tetrabenzyl pyrophosphate
Formula: C₂₈H₂₈O₇P₂
Temperatura de almacenaje: Congelador
Número MDL: MFCD00051941
Núm. CAS: 990-91-0
UN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=93abdcf02ba91ecfe726df85f4ccc917d6472cf9&vid=93abdcf02ba91ecfe726df85f4ccc917d6472cf9&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 60.0 to 64.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR