4-Phenoxypyridine ≥95.0% (by GC, titration analysis)

Proveedor: TCI

P1413-5G
TCIAP1413-5GEA 65 EUR
TCIAP1413-5G
4-Phenoxypyridine ≥95.0% (by GC, titration analysis)
4-Phenoxypyridine
Formula: C₁₁H₉NO
Número MDL: MFCD00235157
Núm. CAS: 4783-86-2

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 95.0 %
Melting point 44.0 to 48.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR