(3-Phenyl-1H-inden-1-ylidene)bis(tricyclohexylphosphine)ruthenium(II) dichloride tetrahydrofuran adduct ≥90.0% (by titrimetric analysis)

Proveedor: TCI
Atención

P1688-100MG
TCIAP1688-100MGEA 28 EUR
TCIAP1688-100MG
(3-Phenyl-1H-inden-1-ylidene)bis(tricyclohexylphosphine)ruthenium(II) dichloride tetrahydrofuran adduct ≥90.0% (by titrimetric analysis)
(3-Phenyl-1H-inden-1-ylidene)bis(tricyclohexylphosphine)ruthenium(II) dichloride tetrahydrofuran
Formula: C₅₁H₇₆Cl₂P₂Ru
Número MDL: MFCD07369034
Núm. CAS: 250220-36-1
EINECS: 480-140-0

Order Now

Resultados test especificaciones

Appearance Amber to Brown powder to crystal
Purity (Argentmetric Titration) min. 90.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR