Probucol ≥98.0% (by HPLC)

Proveedor: TCI

P2002-5G P2002-25G
TCIAP2002-5GEA 39 EUR
TCIAP2002-5G TCIAP2002-25G
Probucol ≥98.0% (by HPLC)
Probucol
Formula: C₃₁H₄₈O₂S₂
Número MDL: MFCD00079281
Núm. CAS: 23288-49-5
EINECS: 245-560-9
Índice Merck: 14,07755
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1569dfd3b875f0cfb98ff4bb00d89a8995b8524a&vid=1569dfd3b875f0cfb98ff4bb00d89a8995b8524a&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 125.0 to 128.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR